Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1305638

Sigma-Aldrich

ANTI-LC3 (APG8A) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, MAP1 light chain 3-like protein 1, MAP1LC3A, Microtubule-associated proteins 1A/1B light chain 3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

14272 Da

species reactivity

human

technique(s)

immunofluorescence: 1:200
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The gene autophagy-related protein 8A (APG8A), also referred to as MAP1LC3A (microtubule associated protein 1 light chain 3 α), is a homologue of Apg8p necessary for autophagy in yeast. The cytosolic form of the protein, upon processing, gets localized to the autophagosome membranes. The LC3 gene is mapped to human chromosome 20q11.22.

Biochem/physiol Actions

The gene MAP1LC3A (microtubule associated protein 1 light chain 3 α) or LC3 or autophagy-related protein 8A (APG8A) encodes a protein that serves as a marker of autophagosomes. It is found to be expressed on surviving oligodendrocytes in Nasu-Hakola disease brains and may be involved in the induction of oligodendrogliopathy.

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ITCH is a putative target for a novel 20q11.22 amplification detected in anaplastic thyroid carcinoma cells by array-based comparative genomic hybridization.
Ishihara T, et al.
Cancer Science (2008)
LC3, an autophagosome marker, is expressed on oligodendrocytes in Nasu-Hakola disease brains.
Satoh J, et al.
Orphanet Journal of Rare Diseases (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service