Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300666

Sigma-Aldrich

Anti-SULT1B1 (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-1B, Anti-Cytosolic, Anti-Member 1, Anti-Sulfotransferase family, Sulfotransferase 1B2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SULT1B1(27284)

Related Categories

General description

Sulfotransferases such as SULT1B1 catalyze the biotransformation of a large number of endogenous compounds such as neurotransmitters, steroids, bile acids, and thyroid hormones, as well as drugs and xenobiotics. Fujita et al. (1997) demonstrated that recombinant rat and human SULT1B1, expressed in E. coli, catalyzed sulfation of p-nitrophenol, 3,3-prime,5-triiodothyronine (T3), and dopamine, but not of beta-estradiol and dehydroepiandrosterone. SULT1B1 showed higher affinities for formation of T3 sulfate than did the phenol sulfotransferases ST1A3 (SULT1A1) or ST1A5. Wang et al. (1998) found that bacterially-expressed SULT1B1 sulfated small phenols such as 1-naphthol and p-nitrophenol and thyroid hormones, including 3,3-prime-diiodothyronine, triiodothyronine, reverse triiodothyronine, and thyroxine. No activity was detected against several sterols or dopamine.

Immunogen

SULT1B1 (NP_055280, 268-306)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human SULT1B1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service