Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1100901

Sigma-Aldrich

Anti-ARPC4 (26-40) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-ARC20, Anti-Actin related protein 2/3 complex, subunit 4, Anti-p20-Arc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~20 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARPC4(10093)

General description

Actin related protein 2/3 complex, subunit 4 (ARPC4) is mapped to human chromosome 3p25.3. It is part of the ARP2/3 complex which connects this complex to the mother filament.

Immunogen

synthetic peptide corresponding to amino acids 26-40 of human ARPC4

Application

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

Actin related protein 2/3 complex, subunit 4 (ARPC4) in plants is crucial for action dynamics and cordinates the tip growth and movement. It is highly expressed in most of the pancreatic carcinoma and may regulate cancer cell migration. It interacts with mycobacterial protein Rv1626 and its expression in Mycobacterium tuberculosis impacts its survival and infection profile, making it an effective anti-tuberculosis drug candidate.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of ARPC4 in tip growth and alignment of the polar axis in filaments of Physcomitrella patens
Perroud PF and Quatrano RS
Cell Motility and the Cytoskeleton, 63(3), 162-171 (2006)
Expression of the ARPC4 subunit of human Arp2/3 severely affects Mycobacterium tuberculosis growth and suppresses immunogenic response in murine macrophages
Ghosh A, et al.
Testing, 8(7), e69949-e69949 (2013)
Silencing of the ARP2/3 complex disturbs pancreatic cancer cell migration
Rauhala HE, et al.
Anticancer Research, 33(1), 45-52 (2013)
3p- syndrome defines a hearing loss locus in 3p25.3
McCullough BJ, et al.
Hearing Research, 224(1-2), 51-60 (2007)
Chiara Galloni et al.
The Journal of cell biology, 220(8) (2021-06-10)
The mechanisms regulating the disassembly of branched actin networks formed by the Arp2/3 complex still remain to be fully elucidated. In addition, the impact of Arp3 isoforms on the properties of Arp2/3 are also unexplored. We now demonstrate that Arp3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service