Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S9994

Sigma-Aldrich

Recombinant SARS-CoV-2 Spike S1-S2 Chimeric Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

conjugate

His

Quality Level

Assay

95%

form

frozen liquid

mol wt

~134 kDa

purified by

chromatography

species reactivity

SARS coronavirus

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.2 mg/mL protein

technique(s)

ELISA: suitable
lateral flow assay: suitable

storage temp.

−70°C

General description

This Spike glycoprotein ectodomain (spikes comprised of multiple S1 & S2 domains) of Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) is expressed in insect cells. Consists of the whole spike antigen. This construct hosts the epitopes for neutralizing antibodies and additional epitopes which can improve sensitivity. It could be important in the case of variant of concern antibody detection, as the characteristic mutations of the different variants of concern are positioned mainly in the Spike protein.
The recombinant SARS-CoV2-Spike S1-S2 Chimeric protein, S9994 is recommended for IgG detection. For IgM detection, a mixture of nucleocapsid and spike proteins is recommended.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service