Skip to Content
MilliporeSigma
All Photos(1)

Documents

S9822

Sigma-Aldrich

Anti-Sonic Hedgehog peptide (C-terminal) antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Dsh, Anti-Hhg1, Anti-Hx, Anti-Shh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

immunohistochemistry: 15 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Shh(20423)

General description

The Hedgehog (Hh) family of secreted proteins consists of sonic hedgehog (Shh), desert hedgehog (Dhh) and Indian hedgehog (Ihh). The Hh signaling regulates cell migration and tissue construction during embryogenesis and reconstruction in adults. Shh protein is secreted by Purkinje cells in the cerebellum with both autocrine and paracrine functions. The Shh signaling is mediated by the interaction of Shh with transmembrane proteins, Patched and Smoothened. The Shh pathway cross-talks Notch and Wnt pathways and acts via downstream targets such as cyclins, γ catenin, myc and other transcription factors. The Hh family members have multiple roles as morphogens, cell survival factors, setting anterior-posterior axis and digit formation during development of limb and axon guidance factors during the development of brain and nervous system. Hedgehog signaling has been implicated in childhood tumors of pancreas, liver, thyroid, gastrointestinal tract, neuroblastomas and medulloblastomas
Anti-Sonic Hedgehog Peptide (C-terminal) recognizes mouse Sonic Hedgehog C-terminal peptide. This antibody shows less than 5% cross-reactivity with rmDhh C-terminal peptide and less than 1% cross-reactivity with a 6X histidine-tagged rmShh N-terminal peptide (amino acids 25-198).

Immunogen

purified, E. coli-derived recombinant mouse 6× histidine-tagged sonic hedgehog C-terminal peptide.

Application

Anti-Sonic Hedgehog Peptide (C-terminal) can be used for immunoblotting at a working concentration of 0.1-0.2 μg/ml. For immunohistochemistry, a working concentration of 15 μg/ml is recommended.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of Sonic hedgehog signaling pathway in neuroblastoma development
Shahi MH et al
Biol. Med., 1, Rev2-Rev2 (2009)
Gijs R van den Brink
Physiological reviews, 87(4), 1343-1375 (2007-10-12)
The Hedgehog family of secreted morphogenetic proteins acts through a complex evolutionary conserved signaling pathway to regulate patterning events during development and in the adult organism. In this review I discuss the role of Hedgehog signaling in the development, postnatal
Alessia Omenetti et al.
American journal of physiology. Gastrointestinal and liver physiology, 294(3), G595-G598 (2008-01-26)
Hedgehog (Hh) signaling modulates tissue remodeling by controlling the fate of Hh-responsive cells. Healthy adult livers exhibit little Hh activity. However, cells involved in adult liver repair, including myofibroblasts and progenitors, are capable of producing and responding to Hh ligands.
Fergal C Kelleher
Carcinogenesis, 32(4), 445-451 (2010-12-28)
To conduct a systematic review of the role that the hedgehog signaling pathway has in pancreatic cancer tumorigenesis. PubMed search (2000-2010) and literature based references. Firstly, in 2009 a genetic analysis of pancreatic cancers found that a core set of
Shohreh F Farzan et al.
American journal of physiology. Gastrointestinal and liver physiology, 294(4), G844-G849 (2008-02-02)
The Hedgehog (Hh) family of secreted proteins is necessary for aspects of the development and maintenance of the gastrointestinal tract. Hh is thought to function as a morphogen, a mitogen, a cell survival factor, and an axon guidance factor. Given

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service