Skip to Content
MilliporeSigma

S8758

N-Succinyl-Ala-Ala-Phe-7-amido-4-methylcoumarin

protease substrate, ≥98%, powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H32N4O8
CAS Number:
Molecular Weight:
564.59
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

N-Succinyl-Ala-Ala-Phe-7-amido-4-methylcoumarin, protease substrate

Quality Segment

assay

≥98%

form

powder

solubility

50% acetic acid: 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(NC(=O)CCC(O)=O)C(=O)NC(C)C(=O)NC(Cc1ccccc1)C(=O)Nc2ccc3C(C)=CC(=O)Oc3c2

InChI

1S/C29H32N4O8/c1-16-13-26(37)41-23-15-20(9-10-21(16)23)32-29(40)22(14-19-7-5-4-6-8-19)33-28(39)18(3)31-27(38)17(2)30-24(34)11-12-25(35)36/h4-10,13,15,17-18,22H,11-12,14H2,1-3H3,(H,30,34)(H,31,38)(H,32,40)(H,33,39)(H,35,36)

InChI key

HHPVJKZZYOXPLH-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library