Skip to Content
MilliporeSigma

S8398

Shields and Sang M3 Insect Medium

With L-glutamine, without potassium bicarbonate, powder, suitable for insect cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

technique(s)

cell culture | insect: suitable

components

NaHCO3: no
L-glutamine: 0.6 g/L
glucose: 10 g/L (Dextro)

shipped in

ambient

storage temp.

2-8°C

General description

Shields and Sang M3 insect medium can support the rapid growth of both primary and established cultures of Drosophila melanogaster cells. Insect tissue culture media have been formulated to mimic the main properties of the hemolymph of specific insects.

Application

Developed for the culture of Drosophila cell lines.
Shields and Sang M3 Insect Medium has been used:
  • for the dissection of Drosophila larvae to study the changes in the translational activity of larval wing imaginal disc cells
  • to incubate Drosophila bodies for surface sensing of translation (SUnSET) assay
  • to culture Drosophila cells for transfection studies



Still not finding the right product?

Explore all of our products under Shields and Sang M3 Insect Medium


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library