Skip to Content
MilliporeSigma
All Photos(1)

Documents

S8197

Sigma-Aldrich

SMER28

>99% (HPLC), solid

Synonym(s):

6-Bromo-N-2-propenyl-4-quinazolinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10BrN3
CAS Number:
Molecular Weight:
264.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

>99% (HPLC)

form

solid

solubility

DMSO: >20 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

Brc1ccc2ncnc(NCC=C)c2c1

InChI

1S/C11H10BrN3/c1-2-5-13-11-9-6-8(12)3-4-10(9)14-7-15-11/h2-4,6-7H,1,5H2,(H,13,14,15)

InChI key

BCPOLXUSCUFDGE-UHFFFAOYSA-N

Application

SMER28 may be used in mTOR-mediated signaling studies.

Biochem/physiol Actions

SMER28 is a small molecule modulator of mammalian autophagy; enhances A53T alpha-synuclein clearance in PC-12 cells independent of rapamycin treatment; appears to act independent of the mTOR pathway, but combined treatment with saturating rapamycin concentration enhances the effect of either compound alone on A53T alpha-synuclein clearance; autophagy inducers may prove useful in the treatment of neurodegenerative and infectious diseases and cancer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dee Shen et al.
Cell biochemistry and biophysics, 60(3), 173-185 (2010-12-07)
Aggresomes and related inclusion bodies appear to serve as storage depots for misfolded and aggregated proteins within cells, which can potentially be degraded by the autophagy pathway. A homogenous fluorescence-based assay was devised to detect aggregated proteins inside aggresomes and
Ann K Rosenthal et al.
The Journal of biological chemistry, 290(21), 13028-13038 (2015-04-15)
Chondrocyte-derived extracellular organelles known as articular cartilage vesicles (ACVs) participate in non-classical protein secretion, intercellular communication, and pathologic calcification. Factors affecting ACV formation and release remain poorly characterized; although in some cell types, the generation of extracellular vesicles is associated
Prashant Bharadwaj et al.
Neural regeneration research, 15(10), 1931-1936 (2020-04-05)
Multiple lines of evidence show that soluble oligomer forms of amyloid β protein (Aβ42) are the most neurotoxic species in the brain and correlates with the degree of neuronal loss and cognitive deficit in Alzheimer's disease. Although many studies have
Yu Wu et al.
The FEBS journal, 287(15), 3184-3199 (2020-01-05)
The endo-lysosome system is involved in endocytosis, protein sorting, and degradation as well as autophagy. Numerous toxins and pathogens exploit this system to enter host cells and exert their deleterious effects. Modulation of host endo-lysosome pathway may restrict multiple toxins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service