MilliporeSigma
S7063
Greiner Multiwell Strips
size 8 wells , strip, high binding surface, non-sterile
Synonym(s):
high binding strip, Greiner Multiwell Strips, Greiner 762061, treated multiwell strips
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.22
UNSPSC Code:
41122107
Material:
polystyrene
Size:
8 wells , strip
Sterility:
non-sterile
Binding type:
high binding surface
Feature:
lid: no
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polystyrene
sterility
non-sterile
feature
lid: no
packaging
pack of 1 ea
manufacturer/tradename
Greiner 762061
maximum volume
388 μL
size
8 wells , strip
working volume
350 μL
binding type
high binding surface
General description
Both medium-binding and high binding are offered. High binding plates and strips are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding are suggested.
Legal Information
Still not finding the right product?
Explore all of our products under Greiner Multiwell Strips
Choose from one of the most recent versions:
Certificates of Analysis (COA)