Skip to Content
MilliporeSigma
All Photos(1)

Documents

S6701

Sigma-Aldrich

SF-11

≥98% (HPLC)

Synonym(s):

N-(4-Ethoxyphenyl)-4-(hydroxydiphenylmethyl)-1-piperidinecarbothioamide, SID 17507305

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30N2O2S
CAS Number:
Molecular Weight:
446.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

CCOc1ccc(NC(=S)N2CCC(CC2)C(O)(c3ccccc3)c4ccccc4)cc1

InChI

1S/C27H30N2O2S/c1-2-31-25-15-13-24(14-16-25)28-26(32)29-19-17-23(18-20-29)27(30,21-9-5-3-6-10-21)22-11-7-4-8-12-22/h3-16,23,30H,2,17-20H2,1H3,(H,28,32)

InChI key

PMEQBGAGFZDWQX-UHFFFAOYSA-N

Application

SF-11 may be used in NPY Y2 receptor-mediated studies.

Biochem/physiol Actions

Radioligand studies reveal that SF-11 also targets 5-HT2B and dopamine transporter.1
SF-11 is a potent, selective NPY Y2 receptor antagonist; brain-penetrant.

Features and Benefits

This compound is featured on the Neuropeptide Y Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaun P Brothers et al.
Molecular pharmacology, 77(1), 46-57 (2009-10-20)
The role of neuropeptide Y Y2 receptor (Y2R) in human diseases such as obesity, mood disorders, and alcoholism could be better resolved by the use of small-molecule chemical probes that are substantially different from the currently available Y2R antagonist, N-[(1S)-4-[(aminoiminomethyl)amino]-1-[[[2-(3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl)ethyl]amino]carbonyl]butyl]-1-[2-[4-(6,11-dihydro-6-oxo-5H-dibenz[b,e]azepin-11-yl)-1-piperazinyl]-2-oxoethyl]-cyclopentaneacetamide)
Ruixin Chen et al.
Neuropeptides, 80, 102022-102022 (2020-01-29)
In recent years, emerging evidence has illustrated the indispensable role of sympathetic neurotransmitters and their receptors in cartilage mediation. The presence of neuropeptide Y (NPY)-positive sympathetic nerve fibres in cartilage and NPY-secretion function in chondrocytes raises the possibility of NPY

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service