Skip to Content
MilliporeSigma
All Photos(2)

Documents

S6322

Sigma-Aldrich

Sigma Adjuvant System®

oil

Synonym(s):

Adjuvant System, Sigma Adjuvant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

form

oil

Quality Level

storage temp.

2-8°C

Application

Adjuvanticity Assay: EIA titer for test mice is at least 2X greater that that of control mice.

Biochem/physiol Actions

Sigma Adjuvant System is a stable oil-in-water emulsion that may be used as an alternative to the classical Freund′s water-in-oil emulsions. It is derived from bacterial and mycobacterial cell wall components that provide potent stimulus to the immune system.

Physical form

Each vial contains 0.5 mg Monophosphoryl Lipid A (detoxified endotoxin) from Salmonella minnesota and 0.5 mg synthetic Trehalose Dicorynomycolate in 2% oil (squalene)-Tween® 80-water.

Legal Information

Sigma Adjuvant System is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Avi-Hai Hovav et al.
Infection and immunity, 73(1), 250-257 (2004-12-25)
In this study, we examined the immunogenicity and protective efficacy of six immunodominant Mycobacterium tuberculosis recombinant antigens (85B, 38kDa, ESAT-6, CFP21, Mtb8.4, and 16kDa) in a multivalent vaccine preparation (6Ag). Gamma interferon (IFN-gamma) and monophosphoryl lipid A-trehalose dicorynomycolate (Ribi) adjuvant
Déirdre B Ní Eidhin et al.
FEMS immunology and medical microbiology, 52(2), 207-218 (2007-12-21)
Clostridium difficile is the leading cause of infectious antibiotic-associated diarrhoea, particularly among the elderly. Its surface-layer protein (SLP) was tested as a vaccine component in a series of immunization and challenge experiments with Golden Syrian hamsters, combined with different systemic
Nadja Thönes et al.
Journal of virology, 82(11), 5472-5485 (2008-04-04)
Capsomeres are considered to be an alternative to viruslike particle (VLP)-based vaccines as they can be produced in prokaryotic expression systems. So far, no detailed side-by-side comparison of VLPs and capsomeres has been performed. In the present study, we immunized
Christina Schellenbacher et al.
Journal of virology, 83(19), 10085-10095 (2009-07-31)
The amino (N) terminus of the human papillomavirus (HPV) minor capsid protein L2 can induce low-titer, cross-neutralizing antibodies. The aim of this study was to improve immunogenicity of L2 peptides by surface display on highly ordered, self-assembled virus-like particles (VLP)
Gabriel Ozorowski et al.
Journal of virology, 92(13) (2018-04-20)
Native-like, soluble, recombinant SOSIP trimers of various designs and based on several env genes of human immunodeficiency virus type 1 (HIV-1) are being tested as immunogens in different animal models. These experiments almost always involve coformulating the trimers with an

Articles

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Related Content

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service