Skip to Content
MilliporeSigma

Skip To

S6191

Sodium chloride

BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture

Synonym(s):

Halite, NaCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
UNSPSC Code:
12352302
NACRES:
NA.25
PubChem Substance ID:
EC Number:
231-598-3
Beilstein/REAXYS Number:
3534976
MDL number:
Assay:
≥99% (titration)
Grade:
BioPerformance Certified
Form:
powder or crystals
Solubility:
water: soluble 100 mg/mL, clear, colorless
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

BioPerformance Certified

Quality Segment

vapor pressure

1.33 (1.00 mmHg) at 865 °C hPa

assay

≥99% (titration)

form

powder or crystals

technique(s)

cell culture | insect: suitable, cell culture | plant: suitable

impurities

endotoxin, tested

pH

7

mp

801 °C (lit.)

solubility

water: soluble 100 mg/mL, clear, colorless

cation traces

≤5 ppm (Heavy metals by ICP-OES)

foreign activity

DNase, RNase, Protease, none detected

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Application

Sodium chloride is suitable reagent for plant cell culture studies and insect cell culture studies. It is the suitable reagent used to prepare alkaline Congo red solution. It may be employed in the basal medium for tobacco cell cultures to investigate the osmotic adjustment of NaCl-adapted and nonadapted cell lines.

Other Notes

Easily compare specifications for Sodium Chloride products with the Sodium Chloride specification table.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
940526793566746398
description

BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture

description

anhydrous, ≥99.99% (trace metals analysis)

description

anhydrous, free-flowing, Redi-Dri™, ReagentPlus®, ≥99%

description

anhydrous, Redi-Dri™, free-flowing, ACS reagent, ≥99%

form

powder or crystals

form

crystalline powder

form

powder

form

powder

assay

≥99% (titration)

assay

≥99.99% (trace metals analysis)

assay

≥99%

assay

≥99%

solubility

water: soluble 100 mg/mL, clear, colorless

solubility

water: soluble 358 g/L at 20 °C

solubility

-

solubility

-

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

100

grade

BioPerformance Certified

grade

-

grade

anhydrous

grade

anhydrous, ACS reagent

mp

801 °C (lit.)

mp

801 °C

mp

801 °C (lit.)

mp

801 °C (lit.)


Still not finding the right product?

Explore all of our products under Sodium chloride


Storage Class

13 - Non Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters