MilliporeSigma
S4767
Wheaton staining dishes
20 slides, covers only
Synonym(s):
900202
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
MA.01
UNSPSC Code:
41100000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
rectangular , soda-lime glass
packaging
pack of 3 ea
manufacturer/tradename
WHEATON® 900202
dish L × W
95 mm × 76 mm
dish capacity
3 x 1 in. (75 x 25 mm) , 10 single slides, 3 x 1-1/2 in. (75 x 38 mm) , 19 slides arranged alternately straight across and diagonally, 3 x 2 in. (75 x 51 mm) , 20 slides back-to-back of standard size
General description
Microscopy is a method of viewing objects that lie outside the resolution of the human eye and can be examined. The objects to be observed are magnified to such an extent that they can be displayed as images. For sample preparation, the objects to be examined are cut into very thin slices. These sections are then applied to an object slide and covered with a cover glass and appropriate solutions. To view the sections under the microscope, the sections on the slide must be filled in a staining trough and stained with an appropriate staining solution.
Features and Benefits
  • Manufactured from soda-lime glass
  • This 20-slide unit is the standard for manual staining procedures
  • The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back of standard size: 3” x 1” (75 x 25mm), 3” x 1-1/2” (75 x 38mm) and 3” x 2” (75 x 51mm) sizes
  • Approximate inside bottom dimensions (mm): 91 L x 71 W x 60 D
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Still not finding the right product?
Explore all of our products under Wheaton staining dishes
Choose from one of the most recent versions:
Certificates of Analysis (COA)