Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S4326

Sigma-Aldrich

SB-277011-A hydrochloride hydrate

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H30N4O · HCl · xH2O
CAS Number:
Molecular Weight:
475.02 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

O.Cl.O=C(N[C@@H]1CC[C@H](CC1)CCN2CCc3cc(ccc3C2)C#N)c4ccnc5ccccc45

InChI

1S/C28H30N4O.ClH.H2O/c29-18-21-5-8-23-19-32(16-13-22(23)17-21)15-12-20-6-9-24(10-7-20)31-28(33)26-11-14-30-27-4-2-1-3-25(26)27;;/h1-5,8,11,14,17,20,24H,6-7,9-10,12-13,15-16,19H2,(H,31,33);1H;1H2/t20-,24-;;

InChI key

OOWNBQACGIKNGM-ONAIBGCWSA-N

Application

SB-277011-A hydrochloride hydrate has been used:
  • as a dopamine D3 receptor-selective antagonist in rats to test its effect on 4-propyl-9-hydroxynaphthoxazine ([3H]-(+)-PHNO) radiotracer in cerebellum L9/10 and striatum
  • as an acetylcholine inhibitor to test its negative allosteric modulation on Ascaris suum nicotinic acetylcholine receptor (Asu-ACR-16) expressed in Xenopus oocytes and its inhibitory effect on locomotion in Caenorhabditis elegans
  • to test its effect on alcohol consumption in mice

SB-277011-A hydrochloride hydrate may be used in D3 dopamine receptor-mediated cell signaling studies.

Biochem/physiol Actions

SB-277011-A is a potent, selective brain-penetrant D3 dopamine receptor antagonist; 80-100x selective for D3 over D2; pKi = 7.95 for hD3 receptor.
The D3 dopamine receptor antagonist, SB-277011-A inhibits the cocaine-seeking behavior and cocaine-enhances brain stimulation reward in in rats. It decreases drug-seeking behavior and may be useful in treatment of cocaine and methamphetamine addiction.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krista Spiller et al.
Psychopharmacology, 196(4), 533-542 (2007-11-07)
We have previously reported that selective antagonism of brain D3 receptors by SB-277011A or NGB 2904 significantly attenuates cocaine- or nicotine-enhanced brain stimulation reward (BSR). In the present study, we investigated whether the selective D3 receptor antagonists SB-277011A and NGB
Zheng-Xiong Xi et al.
The European journal of neuroscience, 21(12), 3427-3438 (2005-07-20)
In rats, acute administration of SB-277011A, a highly selective dopamine (DA) D(3) receptor antagonist, blocks cocaine-enhanced brain stimulation reward, cocaine-seeking behaviour and reinstatement of cocaine-seeking behaviour. Here, we investigated whether SB-277011A attenuates cocaine reinforcement as assessed by cocaine self-administration under
Fudan Zheng et al.
International journal for parasitology. Drugs and drug resistance, 6(1), 60-73 (2016-04-08)
Soil-transmitted helminth infections in humans and livestock cause significant debility, reduced productivity and economic losses globally. There are a limited number of effective anthelmintic drugs available for treating helminths infections, and their frequent use has led to the development of
Jeremy G Gilbert et al.
Synapse (New York, N.Y.), 57(1), 17-28 (2005-04-29)
Recent studies have shown that the novel dopamine (DA) D3 receptor antagonists SB-277011A and NGB 2904 inhibit cocaine- and/or stress-induced reinstatement of drug-seeking behavior. The present study sought to determine if SB-277011A, NGB 2904, or BP-897 (a mixed D3 agonist/antagonist)
Amine Bahi et al.
Alcoholism, clinical and experimental research, 38(9), 2369-2376 (2014-09-27)
It has been reported that dopamine D3 receptor (D3R) knockout mice display similar ethanol (EtOH) consumption compared to wild types. In addition, studies with D3R pharmacological targeting were inconclusive. In the current study, we used both gain- and loss-of-function approaches

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service