Skip to Content
MilliporeSigma

S4311

L-Serine

98.5-101.0%, suitable for cell culture, non-animal source, meets EP, USP testing specifications

Synonym(s):

L Serine, (S)-2-Amino-3-hydroxypropionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
105.09
NACRES:
NA.26
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
EC Number:
200-274-3
MDL number:
Beilstein/REAXYS Number:
1721404
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%

biological source

non-animal source

Quality Segment

agency

USP/NF, meets EP testing specifications, meets USP testing specifications

assay

98.5-101.0%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

mp

222 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL

application(s)

pharmaceutical (small molecule)

SMILES string

N[C@@H](CO)C(O)=O

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1

InChI key

MTCFGRXMJLQNBG-REOHCLBHSA-N

Application

L-Serine has been used as a supplement in a medium to culture Chinese hamster ovary TF 70R cell line and in mouse embryonic fibroblast (MEF)-media.

Biochem/physiol Actions

Precursor of glycine by serine hydroxymethyltransferase.
L-Serine is a non-essential amino acid that acts as a precursor for nucleotide synthesis. It has roles in the development and function of the central nervous system.


Still not finding the right product?

Explore all of our products under L-Serine


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library