Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S3771

Sigma-Aldrich

Ser-Asp-Gly-Arg-Gly

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H30N8O9
CAS Number:
Molecular Weight:
490.47
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (organic)

Assay

≥90% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

NC(CO)C(=O)NC(CC(O)=O)C(=O)NCC(=O)NC(CCCNC(N)=N)C(=O)NCC(O)=O

InChI

1S/C17H30N8O9/c18-8(7-26)14(32)25-10(4-12(28)29)16(34)22-5-11(27)24-9(2-1-3-21-17(19)20)15(33)23-6-13(30)31/h8-10,26H,1-7,18H2,(H,22,34)(H,23,33)(H,24,27)(H,25,32)(H,28,29)(H,30,31)(H4,19,20,21)

InChI key

UVLWLKCNNYTXDT-UHFFFAOYSA-N

Amino Acid Sequence

Ser-Asp-Gly-Arg-Gly

Application

Ser-Asp-Gly-Arg-Gly has been used as a control peptide which does not affect integrin function, in various studies.

Biochem/physiol Actions

Reverse sequence of Gly-Arg-Gly-Asp-Ser, which does not appear to affect integrin function.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic and functional characterisation of the heparin-binding peptides from human transglutaminase 2
Kaupo Teesalu
Journal of Peptide Science (2012)
Y K Liu et al.
Journal of biochemistry, 121(5), 961-968 (1997-05-01)
The beta peptide (113-125), derived from a conserved sequence of the beta subunit of integrins, was synthesized to investigate its adhesive properties to matrix proteins and the effects on cell adhesion to immobilized fibronectin. In this study, we observed that
Characterizing the resolution and accuracy of a second-generation traveling-wave ion mobility separator for biomolecular ions
Yueyang Zhong
Analyst (2011)
F Soncin
Proceedings of the National Academy of Sciences of the United States of America, 89(6), 2232-2236 (1992-03-15)
When coated on bacteriological plastic at doses greater than or equal to 0.1 microgram/cm2, human and bovine angiogenin support calf pulmonary artery endothelial and Chinese hamster fibroblast cell adhesion and spreading, but do not affect cell adhesion when in solution.
Fibronectin induces ureteric bud cells branching and cellular cord and tubule formation
Peng Ye
Kidney International, 66(4) (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service