Skip to Content
MilliporeSigma

S3387

Sucrose 6′-monophosphate dipotassium salt

~98% (TLC)

Synonym(s):

β-D-Fru-6-P-(2→1)-α-D-Glc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H21O14PK2
CAS Number:
Molecular Weight:
498.46
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~98% (TLC)

Quality Level

form

solid

impurities

3.1% water (Karl Fischer)

color

white

storage temp.

−20°C

SMILES string

[K].OCC1OC(OC2(CO)OC(COP(O)(O)=O)C(O)C2O)C(O)C(O)C1O

InChI

1S/C12H23O14P.K.H/c13-1-4-6(15)8(17)9(18)11(24-4)26-12(3-14)10(19)7(16)5(25-12)2-23-27(20,21)22;;/h4-11,13-19H,1-3H2,(H2,20,21,22);;

InChI key

HTUHVZMDNCZYMG-UHFFFAOYSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
S3387-100MG04061832966243
S3387-25MG04061832966267
S3387-10MG04061836922788