Skip to Content

S3381

Sodium stearate

≥99%

Synonym(s):

Octadecanoic acid sodium salt, Stearic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)16COONa
CAS Number:
Molecular Weight:
306.46
UNSPSC Code:
12352211
NACRES:
NA.25
PubChem Substance ID:
EC Number:
212-490-5
Beilstein/REAXYS Number:
3576813
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant (vegetable)

Quality Segment

assay

≥99%

form

powder

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].CCCCCCCCCCCCCCCCCC([O-])=O

InChI

1S/C18H36O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2-17H2,1H3,(H,19,20);/q;+1/p-1

InChI key

RYYKJJJTJZKILX-UHFFFAOYSA-M

Application


  • Stability test of novel combined formulated dry powder inhalation system containing antibiotic: physical characterization and in vitro-in silico lung deposition results.: This study investigates a novel dry powder inhalation system formulated with an antibiotic and sodium stearate. It explores the physical characterization and lung deposition efficiency, demonstrating the potential for enhanced drug delivery to the lungs (Benke et al., 2019).

  • Protective effect of sodium stearate on the moisture-induced deterioration of hygroscopic spray-dried powders.: This research highlights the role of sodium stearate in protecting hygroscopic spray-dried powders from moisture-induced degradation, which is crucial for maintaining the stability and efficacy of pharmaceutical products (Yu et al., 2018).

  • Dissolution of a poorly water-soluble drug dry coated with magnesium and sodium stearate.: This study examines the use of sodium stearate in enhancing the dissolution rates of poorly water-soluble drugs, which is critical for improving drug bioavailability (Tay et al., 2012).



Still not finding the right product?

Explore all of our products under Sodium stearate


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library