Skip to Content
MilliporeSigma

S2646

Scalpel blades

No. 10

Synonym(s):

Precision blades, Surgical scalpel blades

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.35
UNSPSC Code:
41122407
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

steel (carbon)

sterility

non-sterile

Application

Scalpel blades has been used:
  • in seedling inoculation
  • to dissect adrenal gland for adrenalectomy
  • to cut the polydimethylsiloxane (PDMS) layers for fabrication of the microfluidic device

Packaging

individually wrapped.


Still not finding the right product?

Explore all of our products under Scalpel blades




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library