Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S1942

Sigma-Aldrich

S9 from Liver, Pooled

from Monkey(Cynomolgus spp.), male

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161501

biological source

Monkey (Cynomolgus spp.)

Quality Level

packaging

vial of ~20 mg

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

The hepatic S9 pools from a variety of biological sources represent the post-mitochondrial supernatant fraction from homogenized liver. Known to be a rich source of drug metabolizing enzymes including P-450, these pools are useful in the study of xenobiotic metabolism and drug interactions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service