Skip to Content
MilliporeSigma

S1825

S32826 disodium salt hydrate

≥98% (HPLC)

Synonym(s):

[4-(Tetradecanoylamino)benzyl]phosphonic acid disodium salt hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H34NNa2O4P · xH2O
CAS Number:
Molecular Weight:
441.45 (anhydrous basis)
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

H2O: ≥10 mg/mL

storage temp.

−20°C

SMILES string

[Na+].[Na+].CCCCCCCCCCCCCC(=O)Nc1ccc(CP([O-])([O-])=O)cc1

InChI

1S/C21H36NO4P.2Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-21(23)22-20-16-14-19(15-17-20)18-27(24,25)26;;/h14-17H,2-13,18H2,1H3,(H,22,23)(H2,24,25,26);;/q;2*+1/p-2

InChI key

DGRFALMFDGBLCP-UHFFFAOYSA-L

Application

S32826, a potent inhibitor of autotaxin (ATX), may be used to identify and characterize the lyso-phospholipase D (lysoPLD, ATX) involved in bioactive lyso-phosphatidic acid (LPA) formation. S32826 may be used to help determine the role of ATX in malignant cell processes such as tumorigenesis, invasion, and metastases; cardia bifida and atherosclerosis.

Biochem/physiol Actions

S32826 is a potent inhibitor of autotaxin.
S32826 is a potent inhibitor of autotaxin. Autotaxin is a newly discovered lyso-phospholipase D (lysoPLD). Autotaxin and lyso-phosphatidic acid (LPA) have been associated with early cancer progression and motility of cancer cells as well as with metastasis. Because of localization (adipose tissue), the enzyme might play an important role in diabetes and obesity. Autotaxin might be the only source of LPA. S32826 is the strongest inhibitor of autotaxin reported. The compound shows activity in cellular or ex vivo models.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library