Skip to Content
MilliporeSigma

Skip To

RNB700

GenElute Plasma/Serum RNA Purification Maxi Kit

greener alternative
Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.52
UNSPSC Code:
41105600
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

shipped in

ambient

storage temp.

room temp

General description

Sigma′s GenElute Plasma/Serum RNA Purification Maxi Kit provides a fast, reliable, reproducible and simple procedure for isolating circulating RNA and exosomal RNA from small plasma/serum inputs ranging from 2 mL and up to 5 mL, with various kit formats addressing different plasma/serum input volumes. Purification is based on spin column chromatography that uses Sigma′s proprietary resin separation matrix. The kit is designed to isolate all sizes of circulating RNA, including microRNA, as well as all sizes of exosomal RNA. Sigma′s GenElute Plasma/Serum RNA Purification Maxi Kit provides a clear advantage over other available kits in that they do not require phenol/chloroform or any protease treatments. RNA can be isolated from either fresh or frozen samples using these kits. Moreover, the kits allow the user to elute into a flexible elution volume ranging from 10 μL to 100 μL. Typical yields of free-circulating and exosomal RNA vary depending on the input sample, as the amount of RNA present in plasma and serum will depend upon the health status of the individual. Normally, the RNA yield from plasma or serum RNA is highly variable (ranging from 1 to 100 ng/mL).

  • Isolate all sizes of circulating and exosomal RNA, including microRNA
  • Versatile plasma/serum input ranging from 2 mL to 5 mL
  • No phenol extractions
  • No carrier RNA
  • Concentrate circulating RNA and Exosomal RNA into a flexible elution volume ranging from 50 μL to 100 μL
  • Purify high-quality RNA in 35-40 minutes

Application

  • Reverse transcription qPCR
  • Reverse transcription PCR
  • Northern blotting
  • RNase protection and primer extension
  • Expression array assays
  • miRNA array
  • RNA-Seq
  • Nanostring
  • Fluidigm
  • Droplet/Digital PCR

Other Notes

Preps

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RTN70
sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

greener alternative category

Aligned,

greener alternative category

Aligned,

storage temp.

room temp

storage temp.

15-25°C

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

shipped in

ambient

shipped in

-


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters