Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RES5063G-A7

SAFC

L-Glutamic acid, monosodium salt, Low Endotoxin

Synonym(s):

L-Glutamic acid monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
169.11
EC Number:
UNSPSC Code:
12352209
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin and microbial, tested

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

[Na+].N[C@@H](CCC(=O)O)C(=O)[O-]

InChI

1S/C5H9NO4.Na/c6-3(5(9)10)1-2-4(7)8;/h3H,1-2,6H2,(H,7,8)(H,9,10);/q;+1/p-1/t3-;/m0./s1

InChI key

LPUQAYUQRXPFSQ-DFWYDOINSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Application

L-Glutamic acid is a non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Packaging

Product is available in the following package sizes:
RES5063G-A701X: 100g container
RES5063G-A702X: 1kg container
RES5063G-A704X: 10kg container
RES5063G-A705X: 25kg container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service