Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RES2041D

SAFC

DAB

Synonym(s):

3,3′-Diaminobenzidine, 3,3′,4,4′-Biphenyltetramine, 3,3′,4,4′-Tetraaminobiphenyl, DAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2)2C6H3C6H3(NH2)2
CAS Number:
Molecular Weight:
214.27
Beilstein:
1212988
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.21

Quality Level

mp

175-177 °C (lit.)

SMILES string

NC1=C(N)C=CC(C2=CC(N)=C(N)C=C2)=C1

InChI

1S/C12H14N4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6H,13-16H2

InChI key

HSTOKWSFWGCZMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Peroxidase substrate; reagent for spectrophotometric determination of selenium.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service