Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0653

Sigma-Aldrich

Human CCL26 / C-C Motif Chemokine 26 ELISA Kit

Synonym(s):

Eotaxin-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 28 pg/mL
standard curve range: 28.67-7000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL26(10344)

General description

Chemokine ligand 26/ C-C motif chemokine 26 (CCL26) belongs to the eotaxin family. This enzyme-linked immunosorbent assay (ELISA) kit is an in vitro enzyme-linked immunosorbent assay to quantitively measure the target protein in several biological samples, such as serum, plasma, cell culture supernatants, urine, and/or cell and tissue lysates. The antibody pair provided in this kit recognizes human C-C motif chemokine 26.
The antibody pair provided in this kit recognizes human C-C Motif Chemokine 26.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Chemokine ligand 26/ C-C motif chemokine 26 (CCL26) is an eosinophil-selective chemoattractant that interacts with chemokine receptor 3 (CCR3). It plays a role in the pathogenesis of multiple sclerosis (MS). In the central nervous system (CNS), CCL26 is linked with an acute inflammatory response in experimental autoimmune encephalomyelitis (EAE).

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jifei Shou et al.
Journal of neuroimmunology, 333, 576967-576967 (2019-06-01)
Chemokine ligand 26 (CCL26) is a member of the eotaxin family. It works by interacting exclusively with chemokine receptor 3 (CCR3) and acts as an eosinophil-selective chemoattractant. There is an emerging role for eotaxins in autoimmune diseases. Studies have reported
Abumoslem Rashidi Sharifabadi et al.
Clinical laboratory, 60(1), 93-97 (2014-03-08)
Mycobacterium tuberculosis is a pulmonary pathogen responsible for tuberculosis. Tuberculosis (TB) is characterized histologically by granulomas at the site of disease activity. Primary pathologic feature of TB is formation of a granuloma, and chemokines are known to play an important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service