Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0582

Sigma-Aldrich

Cotton Rat IL4 / Interleukin 4 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

cotton rat

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

shipped in

wet ice

storage temp.

−20°C

General description

The antibody pair provided in this kit recognizes Cotton rat IL-4.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abbas Azadmehr et al.
Daru : journal of Faculty of Pharmacy, Tehran University of Medical Sciences, 22, 63-63 (2014-09-18)
Ziziphora tenuior L. (Kakuti in Persian) is used in traditional medicine for treatment of gastrointestinal disorders as carminative and analgesic plant. The other usages of this plant are included treatment of diarrhea and nausea. Therefore in the present study we
Markus M Heimesaat et al.
PloS one, 9(9), e108389-e108389 (2014-09-23)
The neuropeptide Pituitary adenylate cyclase-activating polypeptide (PACAP) plays pivotal roles in immunity and inflammation. So far, potential immune-modulatory properties of PACAP have not been investigated in experimental ileitis. Mice were perorally infected with Toxoplasma (T.) gondii to induce acute ileitis
Bahaa Kenawy Abuel-Hussien Abdel-Salam
Inflammation, 37(5), 1616-1622 (2014-04-25)
The anti-inflammatory cytokines (interleukin (IL)-4 and IL-10) and the pro-inflammatory cytokines (IL-1β, IL-6, and tumor necroses factor-alpha (TNF-α)) have important functions in wound healing. Thus, the aim of this study was to determine whether dietary supplementation with whey protein could
Mohamed El-Behi et al.
European journal of immunology, 44(10), 3003-3014 (2014-07-30)
IL-17-secreting CD8(+) T cells (Tc17 cells) have been implicated in immunity to infections, cancer, and autoimmune diseases. Thus far, studies on Tc17 cells have primarily investigated their development from naïve precursors, while the biology of committed Tc17 cells has been
Fen He et al.
Journal of neuroimmunology, 271(1-2), 1-7 (2014-04-10)
Patients received cranial-irradiation can be affected with cognitive deficits and decreasing hippocampal neurogenesis. In this work, we characterized the cognitive ability and immune-induced neurogenesis of the pre- and post-treated cranial-irradiated rats with Glatiramer acetate (GA), known as a weak CNS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service