Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0513

Sigma-Aldrich

Human VEGF-C ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type urine
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 15 pg/mL
standard curve range: 13.72-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VEGFC(7424)

General description

The Human VEGF-C (Vascular Endothelial Growth Factor- C) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme- linked immunosorbent assay for the quantitative measurement of human VEGF-C in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human VEGF-C

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mizuki Yanase et al.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology, 43(7), 514-520 (2014-04-26)
Vascular endothelial growth factor (VEGF) family members play a major role in angiogenesis and vascularization. VEGF-A promotes tumor angiogenesis by stimulating the growth of tumor vascular endothelial cells. In addition, VEGF-C has been identified as a potent inducer of lymphangiogenesis
Huan Yang et al.
Annals of clinical and laboratory science, 44(2), 158-166 (2014-05-06)
Toll-like receptors (TLRs) play a key role in cancer metastasis. The biological role of TLR2 in invasion and metastasis in gastric carcinoma cells and gastric carcinoma is not clear; therefore, we aimed to investigate the biological role of TLR2 in
Caiqi Ma et al.
International journal of oncology, 54(2), 572-584 (2018-11-30)
Gastric cancer is one of the most malignant tumor types, and its metastasis is a notable cause of mortality. Among the methods of tumor metastasis, lymphatic metastasis is the predominant one in gastric cancer. A previous study reported that the
Chandrasekharam N Nagineni et al.
Aging and disease, 5(2), 88-100 (2014-04-15)
Age-related macular degeneration (AMD) is a sight threating retinal eye disease that affects millions of aging individuals world-wide. Choroid-retinal pigment epithelium (RPE)-neuroretina axis in the posterior compartment of the eye is the primary site of AMD pathology. There are compelling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service