Skip to Content

RAB0442

Phospho-Stat1 (pTyr701) ELISA Kit

for detection of phospho-stat1 (pTyr701) and pan-stat1 in human or mouse cell and tissue lysates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... STAT1(6772)
mouse ... STAT1(20846)
Species reactivity:
human, mouse
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

species reactivity

human, mouse

technique(s)

ELISA: suitable, capture ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... STAT1(6772)
mouse ... STAT1(20846)

General description

The Phospho-Stat1 (pTyr701) ELISA (Enzyme-Linked Immunosorbent Assay) kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates.

Immunogen

Stat1 (pTyr701) synthetic phosphopeptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)


signalword

Warning

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Corrosion

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library