Skip to Content
MilliporeSigma

RAB0438

Cell-Based ELISA for detecting phospho-STAT1 (pTyr701) in cultured cell lines

96 assays

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
41116158
Gene information:
human ... STAT1(6772)
mouse ... STAT1(20846)
Species reactivity:
human, mouse
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

96 assays

species reactivity

human, mouse

technique(s)

ELISA: suitable, capture ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... STAT1(6772)
mouse ... STAT1(20846)

General description

The Cell-Based Stat1 (pTyr701) ELISA kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cells. It can be used for measuring the relative amount of Stat1 (pTyr701) phosphorylation and screening the effects of various treatments, inhibitors (such as siRNA or chemicals), or activators in cultured human and mouse cell lines.

Immunogen

Stat1 (pTyr701) synthetic phosphopeptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • 5x Blocking Solution (Item F)
    SDS

  • Fixing Solution (Item D)
    SDS

  • HRP-conjugated Anti-Rabbit IgG Concentrate (Item I1)
    SDS

  • HRP-conjugated Anti-Mouse IgG Concentrate (Item I2)
    SDS

  • Uncoated Microplate
    SDS

  • Quenching Solution for Cell-based ELISA Assay (Item E)
    SDS

  • Stop Solution
    SDS

See All (10)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library