Skip to Content

RAB0370

Human MMP-8 ELISA Kit

for serum, plasma, conditioned medium, urine

Synonym(s):

MMP-8, Matrix metalloproteinase-8

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... MMP8(4317)
Input:
sample type plasma
sample type serum
sample type urine
sample type conditioned medium
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type plasma
sample type serum
sample type urine
sample type conditioned medium

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 8.23-6000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP8(4317)

General description

The Human MMP-8 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human MMP-8 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human MMP8

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 5X Assay/Sample Diluent Buffer E (Item E2)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library