Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0364

Sigma-Aldrich

Human MMP-13 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type urine
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 8.23-6000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP13(4322)

General description

The Human MMP-13 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human MMP-13 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human MMP13

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

MMP13 (matrix metalloproteinase 13) functions against different types of ECM (extracellular matrix) components. It gets activated by and activates multiple MMPs and hence, has a central role in the MMP cascade. This protein is frequently expressed in colorectal cancer (CRC) and this expression is linked with poor survival in CRC. MMP13 expression is absent in normal breast tissue, but is present in breast carcinoma suggesting its role in tumorigenesis. The expression of this protein can serve as a marker for tumor invasiveness, as its expression is induced during metastasis of multiple types of cancers, including squamous cell carcinomas of the head and neck and this expression is usually linked to poor prognosis. Missense mutation in this gene results in the autosomal dominant disorder called Missouri type of human spondyloepimetaphyseal dysplasia (SEMDMO), which is characterized by abnormal growth and modeling of vertebrae and long bones.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matrix metalloproteinase 13-containing exosomes promote nasopharyngeal carcinoma metastasis.
You Y
Cancer Science null
Yoko Sawada et al.
Experimental and therapeutic medicine, 14(5), 4561-4571 (2017-11-08)
The aim of the present study was to assess the efficacy and safety of the oral administration of
Wen Li et al.
Biomolecules, 9(3) (2019-03-21)
Matrix metalloproteinases (MMPs) are capable of degrading and modifying most components of the extracellular matrix (ECM) and the basal membrane (BM), and play crucial roles in cancer invasion and metastasis. MMP gene expressions were regulated primarily at the transcriptional level
S-S Lin et al.
Osteoarthritis and cartilage, 27(9), 1372-1381 (2019-05-31)
MicroRNA (miRNA)107 expression is downregulated but high mobility group box 1 (HMGB-1), Toll-like receptors (TLRs), and receptor for advanced glycation end products (RAGE) are upregulated in osteoarthritic (OA) cartilage. We investigated mir-107/HMGB-1 signaling in OA after hyperbaric oxygen (HBO) treatment.
X-L Shui et al.
European review for medical and pharmacological sciences, 21(10), 2329-2337 (2017-06-16)
Rheumatoid arthritis (RA) is one systemic auto-immune disorder featured as chronic synovitis and can destruct joint cartilage. Fibroblast-like synoviocyte (FLS) secretes various factors affecting chondrocyte matrix and degradation. This study thus investigated the effect of interleukin-17A (IL-17A) on FLS and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service