Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0330

Sigma-Aldrich

Human SCF ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type urine
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KITLG(4254)

General description

The Human SCF ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human SCF in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human SCF

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaan Oktay et al.
Hormones & cancer, 11(1), 17-33 (2019-12-21)
Breast cancer is the second leading cause of cancer mortality among women. Mammography and tumor biopsy followed by histopathological analysis are the current methods to diagnose breast cancer. Mammography does not detect all breast tumor subtypes, especially those that arise
Armelle Calipel et al.
Clinical & experimental metastasis, 31(5), 553-564 (2014-03-22)
Imatinib mesylate is used in targeted therapy of cancer to inhibit type III tyrosine kinase receptors, such as KIT and platelet-derived growth factor receptors (PDGFRs). Expression of KIT in uveal melanoma (UM) suggests that this receptor may be the target
Charlotte B Westbury et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 111(2), 206-211 (2014-04-22)
Mast cells are involved in the pathogenesis of radiation fibrosis and may be a therapeutic target. The mechanism of increased mast cell number in relation to acute and late tissue responses in human skin was investigated. Punch biopsies of skin
Lawrence M Knab et al.
Molecular cancer research : MCR, 12(10), 1440-1448 (2014-06-20)
Pancreatic ductal adenocarcinoma (PDAC) is associated with a pronounced fibro-inflammatory stromal reaction that contributes to tumor progression. A critical step in invasion and metastasis is the epithelial-to-mesenchymal transition (EMT), which can be regulated by the Snail family of transcription factors.
Timothy B Oriss et al.
Journal of immunology (Baltimore, Md. : 1950), 192(12), 5471-5475 (2014-05-16)
We reported previously that c-kit ligation by membrane-bound stem cell factor (mSCF) boosts IL-6 production in dendritic cells (DCs) and a Th17-immune response. However, Th17 establishment also requires heterodimeric IL-23, but the mechanisms that regulate IL-23 gene expression in DCs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service