Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0151

Sigma-Aldrich

Cell-Based ELISA for detecting phospho-EGFR

in cultured cell lines, 96 assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

usage

96 assays

species reactivity

human

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cultured cells (96 assays)

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EGFR(1965)

General description

The Cell-Based EGFR (activated) ELISA kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cells. It can be used for measuring the relative amount of EGFR (activated) phosphorylation and screening the effects of various treatments, inhibitors (such as siRNA or chemicals), or activators in cultured human cell lines.

Immunogen

Recombinant EGFR (activated)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABBLOCK5x Blocking Solution (Item F)SDS

  • RABFIX1Fixing Solution (Item D)SDS

  • RABHRP1HRP-conjugated Anti-Rabbit IgG Concentrate (Item I1)SDS

  • RABHRP2HRP-conjugated Anti-Mouse IgG Concentrate (Item I2)SDS

  • RABPLATE1Uncoated MicroplateSDS

  • RABQUENCHQuenching Solution for Cell-based ELISA Assay (Item E)SDS

  • RABSTOP1Stop SolutionSDS

  • RABTMB1TMB Substrate Reagent (Item J)SDS

  • RABWASH120X Wash Buffer Concentrate ASDS

See All (9)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service