Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0141

Sigma-Aldrich

Mouse Decorin ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 4.1-1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Dcn(13179)

General description

The Mouse Decorin ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse Decorin in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse Decorin

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoli Niu et al.
Molecular medicine reports, 18(5), 4516-4522 (2018-09-18)
The protective effect of taurine against inflammation, apoptosis and oxidative stress in traumatic brain injury was investigated in the present study. Taurine is a non‑proteogenic and essential amino acid in animals. It plays a critical nutritional role in brain cell
Dong-Ming Kuang et al.
The Journal of clinical investigation, 124(10), 4657-4667 (2014-09-23)
Classical IL-22-producing T helper cells (Th22 cells) mediate inflammatory responses independently of IFN-γ and IL-17; however, nonclassical Th22 cells have been recently identified and coexpress IFN-γ and/or IL-17 along with IL-22. Little is known about how classical and nonclassical Th22
Sergio Occhipinti et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(11), 2910-2921 (2014-03-29)
Despite the great success of HER2 vaccine strategies in animal models, effective clinical results have not yet been obtained. We studied the feasibility of using DNA coding for chimeric rat/human HER2 as a tool to break the unresponsiveness of T
Zhenhong Guo et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(39), 14199-14204 (2014-09-17)
The reciprocal activation of dendritic cells (DCs) and natural killer cells (NKs) plays a key role in both innate and adaptive immunity. The effect of aging on this cross-talk, a critical step in virus disease control and tumor immunology, has
Agnes D Berendsen et al.
Matrix biology : journal of the International Society for Matrix Biology, 35, 223-231 (2014-01-01)
Matrix proteoglycans such as biglycan (Bgn) dominate skeletal tissue and yet its exact role in regulating bone function is still unclear. In this paper we describe the potential role of (Bgn) in the fracture healing process. We hypothesized that Bgn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service