Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0113

Sigma-Aldrich

Mouse Fractalkine / CX3CL1 ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

Quality Level

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 300 pg/mL
standard curve range: 0.274-200 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Cx3cl1(20312)

General description

The Mouse Fractalkine ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse Fractalkine in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse Fractalkine

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fractalkine promotes human monocyte survival via a reduction in oxidative stress.
White GE et.al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 34(12), 2554-2562 (2014)
Increase of serum fractalkine and fractalkine gene expression levels in sickle cell disease patients.
Unal S et.al.
International Journal of Hematology, 101(2), 114-118 (2015)
Gemma E White et al.
Arteriosclerosis, thrombosis, and vascular biology, 34(12), 2554-2562 (2014-11-02)
The CX3C chemokine fractalkine (CX3CL1) has a critical role in the development of atherogenesis because apolipoprotein-E-deficient mice lacking CX3CL1 or its receptor CX3CR1 develop smaller plaques and polymorphisms in CX3CR1 are associated with altered risk of cardiovascular disease. CX3CR1 is
Selma Unal et al.
International journal of hematology, 101(2), 114-118 (2014-12-07)
In the present study, we examined the role of fractalkine (Fkn), a member of the chemokine family, in the pathogenesis of sickle cell disease (SCD). Eighty-seven children with sickle cell disease and 55 healthy children were enrolled in the study.
Junlan Zhang et al.
The American journal of pathology, 184(6), 1706-1714 (2014-04-16)
Hepatic production and release of endothelin-1 (ET-1) binding to endothelin B (ETB) receptors, overexpressed in the lung microvasculature, is associated with accumulation of pro-angiogenic monocytes and vascular remodeling in experimental hepatopulmonary syndrome (HPS) after common bile duct ligation (CBDL). We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service