Skip to Content

RAB0019

Human Amphiregulin ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

AR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... AREG(374)
Input:
sample type plasma
sample type cell culture supernatant(s)
sample type urine
sample type serum
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type urine
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 16.46-4000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AREG(374)

General description

The Human Amphiregulin (AR) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human AR in serum, plasma, cell culture supernatants and urine.
The AREG (amphiregulin) gene codes for a shedding ectodomain, derived from a transmembrane precursor of 252 amino acids. The encoded protein is released into blood or cellular vicinity through autocrine and paracrine. Areg is predominantly expressed in epithelial, mesenchymal cells and also by active immune cells. The AREG gene is mapped to human chromosome 4q13.3.

Immunogen

Recombinant Human Amphiregulin

Application

Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Human Amphiregulin ELISA Kit has been used to estimate the plasma concentration of amphiregulin by ELISA.

Biochem/physiol Actions

Areg (amphiregulin) stimulates growth-signaling pathways via EGF (epidermal growth factor) receptors. Areg participates in the organogenesis of mammary gland. It is regarded as a pro-oncogenic factor and contributes to tumor progression. Upregulation of the AREG gene is observed in cancer and resists chemotherapy. AREG reduces E-cadherin expression in order to promote invasion and progression of ovarian cancer.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 1X Assay/Sample Diluent Buffer A (Item D1)
    SDS

  • ELISA 5X Assay/Sample Diluent Buffer B (Item E1)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library