Skip to Content
MilliporeSigma
All Photos(1)

Documents

R9133

Sigma-Aldrich

Rabbit serum

Synonym(s):

Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

sterility

non-sterile

contains

preservative

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Rabbit serum is pooled from a normal donor population.

Application

Rabbit serum has been used as a control in:
  • immunohistochemistry (IHC)
  • α-toxin neutralization experiments (1:200)
  • indirect immunocytochemistry
  • immunoprecipitation

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rurika Oka et al.
Genome biology, 18(1), 137-137 (2017-07-25)
While most cells in multicellular organisms carry the same genetic information, in each cell type only a subset of genes is being transcribed. Such differentiation in gene expression depends, for a large part, on the activation and repression of regulatory
Benoit Molinie et al.
Nature methods, 13(8), 692-698 (2016-07-05)
N(6)-Methyladenosine (m(6)A) is a widespread, reversible chemical modification of RNA molecules, implicated in many aspects of RNA metabolism. Little quantitative information exists as to either how many transcript copies of particular genes are m(6)A modified ('m(6)A levels') or the relationship
G Cauffman et al.
Human reproduction (Oxford, England), 24(1), 63-70 (2008-10-01)
During human preimplantation development, early blastomeres are believed to be totipotent. It is likely, however, that blastomeres are allocated to a specific lineage prior to any morphological differentiation. NANOG, SOX2 and SALL4 are transcription factors that play a key role
Jiahn-Chun Wu et al.
Journal of cellular biochemistry, 88(4), 823-835 (2003-02-11)
Gap junctions are intercellular communicating channels responsible for the synchronized activity of cardiomyocytes. Recent studies have shown that the membrane-associated guanylate kinase protein, zonula occludens-1 (ZO-1) can bind to catenins in epithelial cells and act as an adapter for the
I L van Wezel et al.
Biology of reproduction, 55(5), 1003-1011 (1996-11-01)
Classically, primordial follicles have flattened pre-granulosa cells that become cuboidal and divide during follicular activation. This change in shape is thus an index of activation. To examine this and follicular morphology in cattle, ovaries were processed for light and electron

Related Content

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service