Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R7020

Sigma-Aldrich

Transcript RNA Markers 0.2-10 kb

for RNA electrophoresis

Synonym(s):

RNA agarose gel electrophoresis ladder, RNA agarose gel electrophoresis marker, northern blot ladder, northern blot marker, ssRNA ladder, ssRNA marker, tRNA ladder, tRNA marker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105337
NACRES:
NA.25

grade

for molecular biology

Quality Level

form

liquid

usage

25 uses

suitability

suitable for electrophoresis (RNA)

storage temp.

−70°C

General description

Sigma′s Transcript RNA Marker (0.2 – 10 kb) contains 9 RNA transcripts from 200 – 10,000 bases. Two ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose gel.

Application

Suitable for size determination of small RNA using native or denaturing agarose gel electrophoresis.
Transcript RNA Markers 0.2-10 kb has been used as a molecular weight marker for northern blot hybridization.

Components

Sigma′s Transcript RNA Marker (0.2 – 10 kb) is provided in a gel-loading solution with 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.

Other Notes

Contains nine defined RNA transcripts, 200–10,000 nucleotides
For optimal resolution, the recommended agarose gel concentration is 1.0%.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barley lesion mimics, supersusceptible or highly resistant to leaf rust and net blotch.
Wright SA, et al.
Plant Pathology, 62, 982-992 (2013)
Magda Dudek et al.
Nucleic acids research, 48(14), 7786-7800 (2020-06-26)
Marine flavobacteria possess dedicated Polysaccharide Utilization Loci (PULs) enabling efficient degradation of a variety of algal polysaccharides. The expression of these PULs is tightly controlled by the presence of the substrate, yet details on the regulatory mechanisms are still lacking.

Articles

Choose the appropriate markers and ladders for nucleic acid size determination of samples separated by electrophoresis. Determine size of DNA, RNA and PCR-generated fragments using agarose or polyacrylamide gels.

Protocols

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Related Content

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service