Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R6265

Sigma-Aldrich

EcoR I from Escherichia coli BS5

Restriction Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.53

grade

for molecular biology

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Specificity

Recognition sequence: 5′-G/AATTC-3′
Cutting results: a 2-10-fold EcoR I overdigestion of 1 μg λ DNA substrate results in 100% cutting
Heat inactivation: 65 °C for 20 minutes.

Application

EcoRI is a restriction endonuclease that is used in molecular biology applications to cleave DNA at the recognition site 5′-G/AATTC-3′, generating fragments with 5′-cohesive termini.

Other Notes

Supplied with 10x Restriction Enzyme Buffer SH (B3657).
Comment: Avoid suboptimal reaction conditions such as low salt, high pH (>8.0) and high glycerol (>5%) as they will alter EcoRI specificity and precipitate star activity.

Unit Definition

One unit is the enzyme activity that completely cleaves 1 mg of λDNA in 1 hour at 37 °C in a total volume of 50 ml of 1x digestion buffer SH for restriction enzymes. 1 mg pBR322 DNA is digested completely by 2 units of EcoR I.

Physical form

Solution in 10 mM Tris-HCl, pH 7.2 , 1 mM EDTA, 200 mM NaCl , 0.5 mM dithioerythritol, 50% glycerol (v/v), 0.2% Triton X- 100 (v/v), 4°C

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Hedgpeth et al.
Proceedings of the National Academy of Sciences of the United States of America, 69(11), 3448-3452 (1972-11-01)
The sequence of DNA base pairs adjacent to the phosphodiester bonds cleaved by the RI restriction endonuclease in unmodified DNA from coliphage lambda has been determined. The 5'-terminal nucleotide labeled with (32)P and oligonucleotides up to the heptamer were analyzed
Jaroslav Jelinek et al.
Epigenetics, 7(12), 1368-1378 (2012-10-19)
Genome wide analysis of DNA methylation provides important information in a variety of diseases, including cancer. Here, we describe a simple method, Digital Restriction Enzyme Analysis of Methylation (DREAM), based on next generation sequencing analysis of methylation-specific signatures created by
Saqib H Ansari et al.
Journal of pediatric hematology/oncology, 35(4), e153-e156 (2013-02-08)
β-thalassemia is characterized by impaired β-chain synthesis leading to ineffective erythropoiesis, severe anemia, and a need for blood transfusion. Presence of Xmn I polymorphism (-158 C-T nucleotide change) in γ-globin gene is associated with a higher fetal hemoglobin and a
Amal M El-Shehaby et al.
Scandinavian journal of clinical and laboratory investigation, 73(1), 75-81 (2012-12-04)
Left ventricular hypertrophy (LVH) is a common manifestation of cardiovascular disease and has an important prognostic value in patients with end-stage renal disease (ESRD). Vitamin D receptor (VDR) has been intensively investigated, and one of these (BsmI) already has been
C Kessler et al.
Gene, 92(1-2), 1-248 (1990-08-16)
The properties and sources of all known class-I, class-II and class-III restriction endonucleases (ENases) and DNA modification methyltransferases (MTases) are listed and newly subclassified according to their sequence specificity. In addition, the enzymes are distinguished in a novel manner according

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service