Skip to Content
MilliporeSigma

R5151

Four-way microtube rack

green

Synonym(s):

adapable tube rack, tube rack, tube storage

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.35
UNSPSC Code:
41122804
Manufacturer/tradename:
Heathrow Scientific HS29022C, Heathrow Scientific
Feature:
Holds 12 x 15 mL tubes, Holds 32 x 0.5 mL tubes, Holds 32 x 1.5-2 mL tubes, Holds 4 x 50 mL tubes
Material:
polypropylene (heavy duty)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene (heavy duty)

description

Autoclavable

feature

Holds 12 x 15 mL tubes, Holds 32 x 0.5 mL tubes, Holds 32 x 1.5-2 mL tubes, Holds 4 x 50 mL tubes

manufacturer/tradename

Heathrow Scientific HS29022C, Heathrow Scientific

size

174x95x52 mm

color

green

General description

  • Racks feature a unique system of tabs and slots, which facilitate easy assembly and sturdy fit.
  • A four-way microtube rack can be connected end-to-end or side-to-side to create a large rack to fit various benchtop applications.
  • Alphanumeric identification enables quick location and retrieval of tubes.
  • The racks can hold 4 × 50 mL conical tubes, 12 × 15 mL conical tubes, 32 × 1.5 mL microtubes, or 32 × 0.5 mL microtubes.
Racks feature a unique system of tabs and slots, which facilitate easy assembly and sturdy fit. The racks can hold 4 × 50 mL conical tubes, 12 × 15 mL conical tubes, 32 × 1.5 mL microtubes or 32 × 0.5 mL microtubes.


Still not finding the right product?

Explore all of our products under Four-way microtube rack




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library