Skip to Content
MilliporeSigma
All Photos(1)

Documents

R354-05A

Sigma-Aldrich

Rat Aortic Smooth Muscle Cells: RAOSMC, adult

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

rat aorta (healthy adult tunica intima and media)

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

morphology

smooth muscle

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


RAOSMC provide an excellent model system to study all aspects of cardiovascular function and disease, especially those related to mechanisms of hyperplasia and hypertrophy of intimal smooth muscle cells leading to vascular occlusion in atherosclerosis and stent restenosis.

RAOSMC from Cell Applications, Inc. have been utilized in dozens of research studies to:

  • Provide a gold standard control for smooth muscle markers Ͱ-SMA, calponin and caldesmon expression (Imamura, 2010)
  • Elucidate cytokines and growth factors signaling pathways implicated in the molecular regulation of smooth muscle cell proliferation, migration, and overall vascular function (Alexis, 2009; Liu, 2009; Tan, 2009; Ko, 2012; Malabanan, 2012; Santiago, 2012; Newaz, 2013); and demonstrate that peach extract is able to inhibit Ang II-induced intracellular Ca2+ elevation and the generation of ROS, thus showing promise in prevention of cardiovascular diseases (Kono, 2013)
  • Investigate extracellular matrix properties and its role in cardiovascular health, repair of damaged vasculature and successful tissue engineering (Chiang, 2009; Kothapalli, 2009a,b,c)
  • Demonstrate that hyperglycemia-induced elastin degradation products lead to vascular smooth muscle cell calcification and identify periostin, also implicated in mechanical stress and inflammation responses, as an early marker for this process (Shanmugam, 2013; Sinha, 2013; Yamashita, 2013)
  • Develop advanced stent technology, including novel surface materials, and drug delivery systems to prevent restenosis and other occlusive vasculopathies (Taite, 2011; Midwinter, 2012; Li, 2013)


Characterization: positive for smooth muscle cell specific alpha-actin expression.

Cell Line Origin

Aorta

Application

Aortic wall contraction, role of smc under normal conditions, blood pressure, distribution of oxygenated blood through systemic circulation.

Components

Rat Smooth Muscle Cell Basal Medium that contains 10% FBS and 10% DMSO

Quality

Each lot was tested for proper morphology, Population Doublings, Negative for HIV, Hepatitis B, Hepatitis C, mycoplasma, bacteria, and fungi.

Preparation Note

  • 2nd passage, >500,000 cells in Rat Smooth Muscle Cell Basal Medium that contains 10% FBS and 10% DMSO
  • Can be cultured at least 16 doublings

Subculture Routine

Please refer to the RAOSMC Culture Protocol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service