Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R2156

Sigma-Aldrich

Relaxin-1 human

recombinant, expressed in E. coli, >95% (SDS-PAGE), lyophilized powder

Synonym(s):

RLN1, RLXH1, Relaxin 1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.26

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

>95% (SDS-PAGE)

form

lyophilized powder

mol wt

(Contains two disulfide-linked subunits: A chain ~2.8 kDa B chain ~3.3 kDa)

technique(s)

cell culture | mammalian: suitable

suitability

endotoxin tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... RLN1(6013)

General description

Cell signaling hormone that is prominent in the prostate, and belongs to the insulin superfamily

Biochem/physiol Actions

Relaxin 1 may function in regulating blood pressure, controlling heart rate, and releasing oxytocin and vasopressin
Relaxin has been shown to have antifibrotic effects in renal fibroblasts through mechanisms that increase collagenase activity and via TGF-β signaling.

Physical form

Lyophilized powder containing BSA

Reconstitution

In Phosphate buffered saline solution containing at least 0.1% serum albumin. The concentration should be ≥10 μg/mL

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosemary Masterson et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 19(3), 544-552 (2004-02-10)
Renal fibroblasts are important effector cells in tubulointerstitial fibrosis, with experimental antifibrotic strategies focusing on the functional down-regulation of these cells. Several experimental models of fibrosis have provided evidence for the effectiveness of the polypeptide hormone relaxin as a potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service