Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R0283

Sigma-Aldrich

Ropivacaine hydrochloride monohydrate

≥98% (HPLC)

Synonym(s):

(2S)-N-(2,6-dimethylphenyl)-1-propyl-2-piperidinecarboxamide hydrochloride monohydrate, (S)-N-(2,6-dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride monohydrate, Naropin hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26N2O · HCl · H2O
CAS Number:
Molecular Weight:
328.88
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -9 to -12, c = 1 in methanol

storage condition

desiccated

color

white to off-white

solubility

H2O: 5 mg/mL, clear

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

O.Cl.CCCN1CCCC[C@H]1C(=O)Nc2c(C)cccc2C

InChI

1S/C17H26N2O.ClH.H2O/c1-4-11-19-12-6-5-10-15(19)17(20)18-16-13(2)8-7-9-14(16)3;;/h7-9,15H,4-6,10-12H2,1-3H3,(H,18,20);1H;1H2/t15-;;/m0../s1

InChI key

VSHFRHVKMYGBJL-CKUXDGONSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ropivacaine has lidocaine-related structure with tertiary amine side chains. It is a pure S(-)enantiomer with a propyl group on the piperidine nitrogen atom.

Application

Ropivacaine hydrochloride monohydrate has been used as an analyte in tandem mass spectrometry and as an amide-based local anaesthetic to test its effect on breast cancer cell methylation.

Biochem/physiol Actions

Local anaesthetic with less cardiotoxicity than bupivacaine.
Local anaesthetic with less cardiotoxicity than bupivacaine; causes reversible blockade of impulse propagation along nerve fibres by preventing the inward movement of sodium ions through the cell membrane of the nerve fibers.
Ropivacaine less lipophilic as compared with bupivacaine. It is metabolized in the liver by the enzyme cytochrome P4501A2. Ropivacaine elicits less central nervous system (CNS) toxicity and is used as a local anaesthetic during postoperative and labour pain.

Features and Benefits

This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jian Zhang et al.
Scientific reports, 8(1), 14181-14181 (2018-09-23)
The purpose of this study is to explore Dvl3 variants and their interaction with negative life events on MDD susceptibility in a Chinese Han population. Additionally, we also attempted to identify whether there is an association between Dvl3 variants and
Janson J White et al.
American journal of human genetics, 98(3), 553-561 (2016-03-01)
Robinow syndrome is a rare congenital disorder characterized by mesomelic limb shortening, genital hypoplasia, and distinctive facial features. Recent reports have identified, in individuals with dominant Robinow syndrome, a specific type of variant characterized by being uniformly located in the
Yuzhuo Wang et al.
International journal of cancer, 142(8), 1602-1610 (2017-12-02)
Traditional pathway analysis map single nucleotide polymorphisms (SNPs) to genes according to physical position, which lacks sufficient biological bases. Here, we incorporated genetics of gene expression into gene- and pathway-based analysis to identify genes and pathways associated with lung cancer
P Lirk et al.
British journal of anaesthesia, 113 Suppl 1, i32-i38 (2014-06-21)
Lidocaine demethylates deoxyribonucleic acid (DNA) in breast cancer cells. This modification of epigenetic information may be of therapeutic relevance in the perioperative period, because a decrease in methylation can reactivate tumour suppressor genes and inhibit tumour growth. The objectives of
Gaurav Kuthiala et al.
Indian journal of anaesthesia, 55(2), 104-110 (2011-06-30)
Ropivacaine is a long-acting amide local anaesthetic agent and first produced as a pure enantiomer. It produces effects similar to other local anaesthetics via reversible inhibition of sodium ion influx in nerve fibres. Ropivacaine is less lipophilic than bupivacaine and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service