Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

R0155

Sigma-Aldrich

Anti-RanGAP1 (C-terminal) antibody produced in rabbit

enhanced validation

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Ran-GTPase-Activating Protein 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~65 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using HeLa cells
indirect immunofluorescence: suitable
western blot: 1-2 μg/mL using HEK-293T cells transfected with human RanGAP1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RANGAP1(5905)
mouse ... Rangap1(19387)

General description

The unmodified form of RanGAP1 (65 kDa) is exclusively cytoplasmic, whereas the 90 kDa SUMO-1-modified form is associated with the cytoplasmic fibers of the nuclear pore complex (NPC).

Immunogen

synthetic peptide corresponding to amino acids 573-587 located at the C-terminus of human RanGAP1. The sequence is highly conserved (80% identity) in mouse RanGAP1.

Application

Anti-RanGAP1 (C-terminal) antibody produced in rabbit has been used for immunoblotting and listeriolysin O (LLO) purification.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

RanGAP1 (Ran GTPase Activating Protein 1, 65 kDa) is a key regulator of Ran activity, specifically inducing the GTPase activity of Ran. RanGAP1 is conjugated to the small ubiquitin-related modifier protein SUMO-1. The association of RanGAP1 with RanBP2 facilitates nuclear transport.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Listeria monocytogenes impairs SUMOylation for efficient infection
Ribet D, et al.
Nature, 464(7292), 1192-1192 (2010)
The RanGAP1-RanBP2 complex is essential for microtubule-kinetochore interactions in vivo
Joseph J, et al.
Current Biology, 14(7), 611-617 (2004)
David Ribet et al.
Nature, 464(7292), 1192-1195 (2010-04-24)
During infection, pathogenic bacteria manipulate the host cell in various ways to allow their own replication, propagation and escape from host immune responses. Post-translational modifications are unique mechanisms that allow cells to rapidly, locally and specifically modify activity or interactions
Smriti Verma et al.
Molecular and cellular biology, 35(17), 2932-2946 (2015-06-24)
Posttranslational modifications (PTMs) can alter many fundamental properties of a protein. One or combinations of them have been known to regulate the dynamics of many cellular pathways and consequently regulate all vital processes. Understandably, pathogens have evolved sophisticated strategies to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service