Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0316

Sigma-Aldrich

PF-06651600

≥98% (HPLC)

Synonym(s):

1-[(2S,5R)-2-Methyl-5-(7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)-1-piperidinyl]-2-propen-1-one malonate, PF 06651600

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19N5O · C3H4O4
CAS Number:
Molecular Weight:
389.41
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

C[C@@H]1N(C(C=C)=O)C[C@H](NC2=NC=NC3=C2C=CN3)CC1.OC(CC(O)=O)=O

InChI

1S/C15H19N5O.C3H4O4/c1-3-13(21)20-8-11(5-4-10(20)2)19-15-12-6-7-16-14(12)17-9-18-15;4-2(5)1-3(6)7/h3,6-7,9-11H,1,4-5,8H2,2H3,(H2,16,17,18,19);1H2,(H,4,5)(H,6,7)/t10-,11+;/m0./s1

InChI key

QMPMPSGDPRHZCG-VZXYPILPSA-N

Biochem/physiol Actions

PF-06651600 is a potent and selective JAK3 inhibitor.
PF-06651600 specifically targets the ATP (adenosine triphosphate)-binding domain of JAK3 (Janus kinase 3). Loss of function of JAK3 is associated with severe combined immunodeficiency syndrome.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jak3 deficiency blocks innate lymphoid cell development.
Robinette M L, et al.
Mucosal Immunology, 11(1), 50-50 (2018)
Targeting JAK/STAT signalling in inflammatory skin diseases with small molecule inhibitors.
Welsch K, et al.
European Journal of Immunology, 47(7), 1096-1107 (2017)
Zhenpeng Dai et al.
JCI insight, 6(7) (2021-04-09)
The Janus kinase/signal transducers and activators of transcription (JAK/STAT) are key intracellular mediators in the signal transduction of many cytokines and growth factors. Common γ chain cytokines and interferon-γ that use the JAK/STAT pathway to induce biological responses have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service