Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0170

Sigma-Aldrich

Torcetrapib

≥98% (HPLC)

Synonym(s):

(2R,4S)-4-((3,5-Bis-trifluoromethylbenzyl)methoxycarbonylamino)-2-ethyl-6-trifluoromethyl-3,4-dihydro-2H-quinoline-1-carboxylic acid ethyl ester, CP-529,414, CP-529414

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H25F9N2O4
CAS Number:
Molecular Weight:
600.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D >-70°

color

white

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

CCOC(=O)N1[C@H](CC)C[C@H](N(Cc2cc(cc(c2)C(F)(F)F)C(F)(F)F)C(=O)OC)c3cc(ccc13)C(F)(F)F

InChI

1S/C26H25F9N2O4/c1-4-18-12-21(19-11-15(24(27,28)29)6-7-20(19)37(18)23(39)41-5-2)36(22(38)40-3)13-14-8-16(25(30,31)32)10-17(9-14)26(33,34)35/h6-11,18,21H,4-5,12-13H2,1-3H3/t18-,21+/m1/s1

InChI key

CMSGWTNRGKRWGS-NQIIRXRSSA-N

Application

Torcetrapib has been used as a reference standard in the medium chain-lipid based formulations.

Biochem/physiol Actions

Cholesteryl ester transfer protein (CETP) inhibitor.
Torcetrapib is a Cholesteryl ester transfer protein (CETP) inhibitor. CETP normally transfers cholesterol from HDL cholesterol to very low density or low density lipoproteins (VLDL or LDL). Inhibition of this process results in higher HDL levels (the "good" cholesterol-containing particle) and reduces LDL levels (the "bad" cholesterol). Unfortunately clinical trials were stopped because of excessive all cause mortality. Reasons are still being investigated, but may be related to some off target effects such as an increase in aldosterone secretion not found in some other CETP inhibitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengjun Fan et al.
BMC systems biology, 6, 152-152 (2012-12-12)
Torcetrapib, a cholesteryl ester transfer protein (CETP) inhibitor which raises high-density lipoprotein (HDL) cholesterol and reduces low-density lipoprotein (LDL) cholesterol level, has been documented to increase mortality and cardiac events associated with adverse effects. However, it is still unclear the
Anatol Kontush et al.
Nature clinical practice. Cardiovascular medicine, 5(6), 329-336 (2008-04-24)
Subnormal levels of HDL cholesterol constitute a major cardiovascular risk factor. Inhibitors of cholesteryl ester transfer protein (CETP) are presently the most potent HDL-raising agents. Torcetrapib was the first CETP inhibitor to enter a large-scale, prospective, placebo-controlled interventional trial, which
Aldosterone, sodium, and hypertension: lessons from torcetrapib?
John W Funder
Hypertension (Dallas, Tex. : 1979), 55(2), 221-223 (2010-01-06)
Philip J Barter et al.
Journal of lipid research, 53(11), 2436-2442 (2012-09-04)
Development of the cholesteryl ester transfer protein (CETP) inhibitor, torcetrapib, was halted after the ILLUMINATE trial revealed an increase in both all-cause mortality (ACM) and major cardiovascular events (MCVEs) associated with its use. We now report that the harm caused
Bernd Hewing et al.
Current opinion in lipidology, 23(4), 372-376 (2012-04-21)
Raising HDL cholesterol (HDL-C) has become an attractive therapeutic target to lower cardiovascular risk in addition to statins. Inhibition of the cholesteryl ester transfer protein (CETP), which mediates the transfer of cholesteryl esters from HDL to apolipoprotein B-containing particles, leads

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service