Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0036

Sigma-Aldrich

Rucaparib camsylate

≥98% (HPLC)

Synonym(s):

8-fluoro-1,3,4,5-tetrahydro-2-[4-[(methylamino)methyl]phenyl]-6H-Pyrrolo[4,3,2-ef][2]benzazepin-6-one camsylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18FN3O · C10H16O4S
CAS Number:
Molecular Weight:
555.66
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

pale yellow

solubility

DMSO: 2 mg/mL, clear

shipped in

dry ice

storage temp.

−20°C

SMILES string

FC1=CC(C(NCC2)=O)=C3C2=C(C4=CC=C(CNC)C=C4)NC3=C1.CC5(C)[C@@H]6CC[C@@]5(CS(O)(=O)=O)C(C6)=O

InChI

1S/C19H18FN3O.C10H16O4S/c1-21-10-11-2-4-12(5-3-11)18-14-6-7-22-19(24)15-8-13(20)9-16(23-18)17(14)15;1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h2-5,8-9,21,23H,6-7,10H2,1H3,(H,22,24);7H,3-6H2,1-2H3,(H,12,13,14)/t;7-,10-/m.1/s1

InChI key

INBJJAFXHQQSRW-STOWLHSFSA-N

Biochem/physiol Actions

PARP Inhibitor; anticancer agent
Rucaparib is an inhibitor of the DNA repair enzyme poly-ADP ribose polymerase-1 (PARP-1). It has been found to have anticancer activity in a number of cancers and has been approved for treatment of previously treated, BRCA-mutant ovarian cancer.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaroslava Karpova et al.
Biochemical pharmacology, 167, 149-162 (2019-03-19)
In our previous studies of the molecular mechanisms of poly(ADP-ribose) polymerase 1 (PARP-1)-mediated transcriptional regulation we identified a novel class of PARP-1 inhibitors targeting the histone-dependent route of PARP-1 activation. Because histone-dependent activation is unique to PARP-1, non-NAD-like PARP-1 inhibitors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service