Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0031

Sigma-Aldrich

PF-03882845

≥98% (HPLC)

Synonym(s):

(3S,3aR)-2-(3-Chloro-4-cyanophenyl)-3-cyclopentyl-3,3a,4,5-tetrahydro-2H-benzo[g]indazole-7-carboxylic acid, PF-3882845

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22ClN3O2
CAS Number:
Molecular Weight:
419.90
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

OC(C1=CC2=C(C=C1)C3=NN(C4=CC=C(C#N)C(Cl)=C4)[C@@H](C5CCCC5)[C@@]3([H])CC2)=O

InChI

1S/C24H22ClN3O2/c25-21-12-18(8-5-17(21)13-26)28-23(14-3-1-2-4-14)20-10-6-15-11-16(24(29)30)7-9-19(15)22(20)27-28/h5,7-9,11-12,14,20,23H,1-4,6,10H2,(H,29,30)/t20-,23-/m0/s1

InChI key

XNULRSOGWPFPBL-REWPJTCUSA-N

Biochem/physiol Actions

PF-03882845 is a potent non-steroidal mineralocorticoid antagonist with and IC50 of 0.755 nM, compared to eplerenone with an IC50 of 109 nM. PF-03882845 was tested in a rat model for renal protection against aldosterone-mediated renal disease, and found to be more potent than eplerenone in suppressing the urinary albumin to creatinine ratio (UACR), a measure of renal fibrosis. The therapeutic index, calculated as the ratio of the EC50 for increasing serum K+ to the EC50 for UACR lowering, was 83.8 for PF-03882845 and 1.47 for eplerenone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service