Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PRS3969

Sigma-Aldrich

Anti-Sumo antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GAP-modifying protein 1, Anti-SMT3 suppressor of mif two 3 homolog 1 (S. cerevisiae), Anti-Ubiquitin-like protein 1, Anti-small-ubiquitin-like modifier 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SUMO1(7341)

General description

SUMO1 (small ubiquitin-related modifier) is also called as Pic1 (inner centromere protein-related protein), Ubl1 (ubiquitin-like protein), hSmt3 (human homolog 3), or sentrin. It is located in the nucleus. SUMO1 gene is located on human chromosome 2q33.

Immunogen

a 14 amino acid peptide from near the amino-terminus of human sumo.

Biochem/physiol Actions

Suppressing SUMO1 (small ubiquitin-related modifier) stops multiplication and induces apoptosis of gastric cancer SGC-7901 cells. It is essential for HCV (hepatitis C virus) replication. Deletion of SUMO1 blocks the aggregation of lipid droplets. It involves in mitochondrial fission.
Suppressing SUMO1 (small ubiquitin-related modifier) stops multiplication and induces apoptosis of gastric cancer SGC-7901 cells. It is essential for HCV (hepatitis C virus) replication. Deletion of SUMO1 blocks the aggregation of lipid droplets. It is involved in mitochondrial fission.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3969.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The genetic basis of cleft lip and cleft palate
Veros C, et al.
Aristotle University Medical Journal, 43 (2016)
Sumo1 conjugates mitochondrial substrates and participates in mitochondrial fission
Harder Z, et al.
Current Biology, 14(4), 340-345 (2004)
The nucleoporin RanBP2 has SUMO1 E3 ligase activity
Pichler A, et al.
Cell, 108(1), 109-120 (2002)
SUMO1 depletion prevents lipid droplet accumulation and HCV replication
Akil A, et al.
Archives of Virology, 161(1), 141-148 (2016)
SUMO-1 Gene Silencing Inhibits Proliferation and Promotes Apoptosis of Human Gastric Cancer SGC-7901 Cells
Jin L, et al.
Cellular Physiology and Biochemistry, 41(3), 987-998 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service