Skip to Content

PLA0154

Rabbit anti-PPP1CA Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

PP-1A, PP1A, PP1alpha, PPP1A, alpha isoform, alpha isozyme, catalytic subunit, protein phosphatase 1, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, testicular tissue protein Li 155

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:500- 1:2,000, western blot: 1:2,000- 1:10,000

accession no.

NP_002699.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... PPP1CA(5499)

Immunogen

The epitope recognized by PLA0154 maps to a region between residue 280 and the C-terminus (residue 330) of human Protein Phosphatase 1, Catalytic Subunit, Alpha Isoform using the numbering given in entry NP_002699.1 (GeneID 5499).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Protein phosphatase 1 (PP1) is a major eukaryotic serine/threonine phosphatase that is involved in a multitude of cellular functions. PP1 is a holoenzyme that consists of the PP1 catalytic subunit (PP1c) that associates with and is regulated by over 50 regulatory subunits. There are several gene and alternative splice products of the catalytic subunit PP1c. PPP1CA encodes the PP1 alpha catalytic subunit. In addition to PP1 alpha, there is PP1 beta (PPP1CB), and two splice variants of PP1 gamma (PPP1CC). The mutually exclusive interaction between the various catalytic and regulatory subunits allows the PP1 enzyme to be involved in a variety of functions which include cell division, glycogen metabolism, muscle contractility, and protein synthesis. serine/threonine-protein phosphatase PP1-alpha catalytic subunit, PPP1A, PP-1A, MGC1674, MGC15877.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library